accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 199287-52-0

IUPAC Name: 4,5-difluoro-2-hydroxybenzaldehyde
Synonyms: 4,5-difluoro-2-hydroxybenzaldehyde
Molecular Weight (g/mol): 158.1
Molecular Formula: C7H4F2O2
InChi Key: HRLWPXGNZQEUHM-UHFFFAOYSA-N
SMILES: OC1=CC(F)=C(F)C=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results