accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 199296-40-7

H3CH3CCH3OONHNO

CAS RN 199296-40-7

IUPAC Name: tert-butyl N-(5-formylpyridin-2-yl)carbamate
Synonyms: tert-butyl N-(5-formylpyridin-2-yl)carbamate
Molecular Weight (g/mol): 222.24
Molecular Formula: C11H14N2O3
InChi Key: WZROBBWIJBBWQP-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=NC=C(C=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results