accessibility menu, dialog, popup

UserName

Viewing profile as user:
OONH2SSOONH2H2ONaNa

CAS RN 199329-53-8

IUPAC Name: disodium 2-amino-3-[(2-amino-2-carboxylatoethyl)disulfanyl]propanoate hydrate
Synonyms: disodium 2-amino-3-[(2-amino-2-carboxylatoethyl)disulfanyl]propanoate hydrate
Molecular Weight (g/mol): 302.27
Molecular Formula: C6H12N2Na2O5S2
InChi Key: TVPDKJXPEMKMDM-UHFFFAOYNA-L
SMILES: O.[Na+].[Na+].NC(CSSCC(N)C([O-])=O)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)