CAS RN 1994-13-4
IUPAC Name:
6-fluoro-4-hydroxy-2H-chromen-2-one
Synonyms:
6-fluoro-4-hydroxychromen-2-one
Molecular Weight (g/mol):
180.13
Molecular Formula:
C9H5FO3
InChi Key:
JCOYNGQUPPGRNB-UHFFFAOYSA-N
SMILES:
OC1=CC(=O)OC2=CC=C(F)C=C12
1
–
1
of
1
results
1
6-Fluoro-4-hydroxycoumarin 98.0+%, TCI America™
CAS: 1994-13-4 Molecular Formula: C9H5FO3 Molecular Weight (g/mol): 180.13 MDL Number: MFCD03094006 InChI Key: JCOYNGQUPPGRNB-UHFFFAOYSA-N PubChem CID: 54691406 IUPAC Name: 6-fluoro-4-hydroxy-2H-chromen-2-one SMILES: OC1=CC(=O)OC2=CC=C(F)C=C12