accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19947-75-2

IUPAC Name: dimethyl-1,2-oxazol-5-amine
Synonyms: dimethyl-1,2-oxazol-5-amine
Molecular Weight (g/mol): 112.13
Molecular Formula: C5H8N2O
InChi Key: PYNDWPFZDQONDV-UHFFFAOYSA-N
SMILES: CC1=NOC(N)=C1C
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results