accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 199526-97-1

IUPAC Name: 4,6-difluoro-1H-indole
Synonyms: 4,6-difluoro-1H-indole
Molecular Weight (g/mol): 153.13
Molecular Formula: C8H5F2N
InChi Key: MHICCULQVCEWFH-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C2C=CNC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results