accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 199541-17-8

H3CNHCH3PPPPH2ClH2ClH2ClHClClRuRu(?)(?)

CAS RN 199541-17-8

IUPAC Name: trichloranium bis([2'-(diphenylphosphanyl)-[1,1'-binaphthalen]-2-yl]diphenylphosphane) dimethylamine hydrochloride diruthenium chloride
Synonyms: trichloranium bis(binap) dimethylamine hydrochloride diruthenium chloride
Molecular Weight (g/mol): 1676.9
Molecular Formula: C90H78Cl5NP4Ru2
InChi Key: IDNUVQHPQCZKMV-UHFFFAOYSA-M
SMILES: Cl.[ClH2+].[ClH2+].[ClH2+].[Cl-].[Ru].[Ru].CNC.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results