accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 199609-62-6

IUPAC Name: [3-({[(tert-butoxy)carbonyl]amino}methyl)phenyl]boronic acid
Synonyms: 3-{[(tert-butoxycarbonyl)amino]methyl}phenylboronic acid
Molecular Weight (g/mol): 251.09
Molecular Formula: C12H18BNO4
InChi Key: YHAQUGOSDQZIMA-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCC1=CC=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results