accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19962-06-2

IUPAC Name: tert-butyl N-(3-hydroxyphenyl)carbamate
Synonyms: tert-butyl N-(3-hydroxyphenyl)carbamate
Molecular Weight (g/mol): 209.25
Molecular Formula: C11H15NO3
InChi Key: HJQNVUQTARSZDK-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=CC=CC(O)=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results