accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 199684-47-4

IUPAC Name: bis([2'-(diphenylphosphanyl)-[1,1'-binaphthalen]-2-yl]diphenylphosphane) [chloraniumyl(chloro)ruthenio]chloranium dimethylamine hydrochloride ruthenium chloride
Synonyms: [chloraniumyl(chloro)ruthenio]chloranium bis(binap) dimethylamine hydrochloride ruthenium chloride
Molecular Weight (g/mol): 1673.88
Molecular Formula: C90H75Cl5NP4Ru2
InChi Key: NVDGOPZSESFSJS-UHFFFAOYSA-M
SMILES: Cl.[Cl-].[Ru].CNC.[ClH+][Ru-]([ClH+])[ClH+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results