accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSiH3CCH3O

CAS RN 19980-43-9

IUPAC Name: (cyclopent-1-en-1-yloxy)trimethylsilane
Synonyms: (cyclopent-1-en-1-yloxy)trimethylsilane
Molecular Weight (g/mol): 156.3
Molecular Formula: C8H16OSi
InChi Key: UBMYYGXGMPGCBO-UHFFFAOYSA-N
SMILES: C[Si](C)(C)OC1=CCCC1
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results