accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 199866-90-5

IUPAC Name: 1,4-difluoro-2,5-dimethoxybenzene
Synonyms: 1,4-difluoro-2,5-dimethoxybenzene
Molecular Weight (g/mol): 174.15
Molecular Formula: C8H8F2O2
InChi Key: QNQUBUBFPGHXAL-UHFFFAOYSA-N
SMILES: COC1=CC(F)=C(OC)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results