accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSNHOCH3NH2OHO

CAS RN 1999-43-5

IUPAC Name: 2-(2-aminopropanamido)-4-(methylsulfanyl)butanoic acid
Synonyms: 2-(2-aminopropanamido)-4-(methylsulfanyl)butanoic acid
Molecular Weight (g/mol): 220.29
Molecular Formula: C8H16N2O3S
InChi Key: FSHURBQASBLAPO-UHFFFAOYNA-N
SMILES: CSCCC(NC(=O)C(C)N)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)