accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CHNOH3CH2NOHO

CAS RN 1999-46-8

IUPAC Name: 2-(2-aminopropanamido)-3-methylbutanoic acid
Synonyms: 2-(2-aminopropanamido)-3-methylbutanoic acid
Molecular Weight (g/mol): 188.23
Molecular Formula: C8H16N2O3
InChi Key: LIWMQSWFLXEGMA-UHFFFAOYNA-N
SMILES: CC(C)C(NC(=O)C(C)N)C(O)=O
Grade 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results