accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20012-63-9

NH2O

CAS RN 20012-63-9

IUPAC Name: 2-(benzyloxy)aniline
Synonyms: 2-(benzyloxy)aniline
Molecular Weight (g/mol): 199.25
Molecular Formula: C13H13NO
InChi Key: PLPVLSBYYOWFKM-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1OCC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results