accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHHO

CAS RN 20016-85-7

IUPAC Name: 2-hydroxybutanoic acid
Synonyms: α-hydroxybutyric acid (+/-)α-hydoxy butyric acid
Molecular Weight (g/mol): 104.11
Molecular Formula: C4H8O3
InChi Key: AFENDNXGAFYKQO-UHFFFAOYNA-N
SMILES: CCC(O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results