accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOO

CAS RN 20029-52-1

IUPAC Name: 4-cyclohexylbenzoic acid
Synonyms: 4-cyclohexylbenzoic acid
Molecular Weight (g/mol): 204.27
Molecular Formula: C13H16O2
InChi Key: QCIWHVKGVVQHIY-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)C1CCCCC1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)