accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOONHO

CAS RN 2003-79-4

IUPAC Name: 2-oxo-1,2-dihydroquinoline-3-carboxylic acid
Synonyms: 2-oxo-1H-quinoline-3-carboxylic acid
Molecular Weight (g/mol): 189.17
Molecular Formula: C10H7NO3
InChi Key: XOQQVKDBGLYPGH-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC2=CC=CC=C2NC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results