accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSOHONHOO

CAS RN 200354-34-3

IUPAC Name: 3-(ethylsulfanyl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms: 3-(ethylsulfanyl)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 371.45
Molecular Formula: C20H21NO4S
InChi Key: OUXUHWHQQLGDQE-UHFFFAOYNA-N
SMILES: CCSCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)