accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20039-37-6

IUPAC Name: [(hydroxydioxochromio)oxy]chromiumoylol; bis(pyridine)
Synonyms: cornforth reagent
Molecular Weight (g/mol): 376.21
Molecular Formula: C10H12Cr2N2O7
InChi Key: RCBVKBFIWMOMHF-UHFFFAOYSA-L
SMILES: C1=CC=NC=C1.C1=CC=NC=C1.O[Cr](=O)(=O)O[Cr](O)(=O)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (5)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (6)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (6)
  • (3)
  • (2)
  • (5)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (6)
  • (5)