accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3COOOHONCH3OO

CAS RN 200616-40-6

IUPAC Name: 5-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}(methyl)amino)-5-oxopentanoic acid
Synonyms: 5-(tert-butoxy)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl](methyl)amino}-5-oxopentanoic acid
Molecular Weight (g/mol): 439.51
Molecular Formula: C25H29NO6
InChi Key: FVUASVBQADLDRO-UHFFFAOYNA-N
SMILES: CN(C(CCC(=O)OC(C)(C)C)C(O)=O)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)