accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COPONH2OCH3

CAS RN 20074-79-7

IUPAC Name: diethyl [(4-aminophenyl)methyl]phosphonate
Synonyms: diethyl (4-aminophenyl)methylphosphonate
Molecular Weight (g/mol): 243.24
Molecular Formula: C11H18NO3P
InChi Key: ZVAYUUUQOCPZCZ-UHFFFAOYSA-N
SMILES: CCOP(=O)(CC1=CC=C(N)C=C1)OCC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results