accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNClClCl

CAS RN 200807-45-0

IUPAC Name: 2-chloro-N-(2,6-dichlorophenyl)benzamide
Synonyms: 2-chloro-N-(2,6-dichlorophenyl)benzamide
Molecular Weight (g/mol): 300.56
Molecular Formula: C13H8Cl3NO
InChi Key: MLILWLKTWNAOFL-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(Cl)=C1NC(=O)C1=CC=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results