accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClHNOClCl

CAS RN 200807-46-1

IUPAC Name: 4-chloro-N-(2,6-dichlorophenyl)benzamide
Synonyms: 4-chloro-N-(2,6-dichlorophenyl)benzamide
Molecular Weight (g/mol): 300.56
Molecular Formula: C13H8Cl3NO
InChi Key: MGCCUXCCTBKJAN-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C(=O)NC1=C(Cl)C=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results