CAS RN 20083-34-5
IUPAC Name:
triethoxy(2,3,4,5,6-pentafluorophenyl)silane
Synonyms:
triethoxy(2,3,4,5,6-pentafluorophenyl)silane
Molecular Weight (g/mol):
330.33
Molecular Formula:
C12H15F5O3Si
InChi Key:
QALDFNLNVLQDSP-UHFFFAOYSA-N
SMILES:
CCO[Si](OCC)(OCC)C1=C(F)C(F)=C(F)C(F)=C1F
1
–
1
of
1
results
1
Triethoxy(pentafluorophenyl)silane 95.0+%, TCI America™
CAS: 20083-34-5 Molecular Formula: C12H15F5O3Si Molecular Weight (g/mol): 330.326 MDL Number: MFCD03411257 InChI Key: QALDFNLNVLQDSP-UHFFFAOYSA-N PubChem CID: 2782625 IUPAC Name: triethoxy-(2,3,4,5,6-pentafluorophenyl)silane SMILES: CCO[Si](C1=C(C(=C(C(=C1F)F)F)F)F)(OCC)OCC