accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 200880-40-6

IUPAC Name: sodium 1-[(2,3-dihydroxypropyl phosphonato)oxy]-3-(tetradecanoyloxy)propan-2-yl tetradecanoate
Synonyms: sodium 1-[(2,3-dihydroxypropyl phosphonato)oxy]-3-(tetradecanoyloxy)propan-2-yl tetradecanoate
Molecular Weight (g/mol): 688.86
Molecular Formula: C34H66NaO10P
InChi Key: QLNOOKSBAYIHQI-UHFFFAOYNA-M
SMILES: [Na+].CCCCCCCCCCCCCC(=O)OCC(COP([O-])(=O)OCC(O)CO)OC(=O)CCCCCCCCCCCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results