accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOCl

CAS RN 2009-83-8

IUPAC Name: 6-chlorohexan-1-ol
Synonyms: 1-hexanol, 6-chloro-
Molecular Weight (g/mol): 136.62
Molecular Formula: C6H13ClO
InChi Key: JNTPTNNCGDAGEJ-UHFFFAOYSA-N
SMILES: OCCCCCCCl
Boiling Point 
  • (2)
  • (3)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (2)
  • (3)
Quantity 
  • (2)
  • (4)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (6)
  • (2)
  • (3)
  • (3)
  • (2)
  • (5)
  • (5)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (4)
  • (1)
  • (3)
  • (5)