CAS RN 20098-20-8
IUPAC Name:
5-bromoadamantan-2-one
Synonyms:
5-bromoadamantan-2-one
Molecular Weight (g/mol):
229.12
Molecular Formula:
C10H13BrO
InChi Key:
TXEWIOREYPSNRR-UHFFFAOYNA-N
SMILES:
BrC12CC3CC(C1)C(=O)C(C3)C2
1
–
1
of
1
results
1
5-Bromo-2-adamantanone 98.0+%, TCI America™
CAS: 20098-20-8 Molecular Formula: C10H13BrO Molecular Weight (g/mol): 229.117 MDL Number: MFCD01321097 InChI Key: TXEWIOREYPSNRR-UHFFFAOYSA-N PubChem CID: 590905 IUPAC Name: 5-bromoadamantan-2-one SMILES: C1C2CC3CC(C2)(CC1C3=O)Br