accessibility menu, dialog, popup

UserName

Viewing profile as user:
(S)(R)(R)O(R)(S)(R)(R)O(R)NONH2HOOHOPOOOOHHONNNNNH2OPOONaH2OH2O

CAS RN 20111-18-6

IUPAC Name: sodium 1-[(2R,3R,4S,5R)-5-({[({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)(oxido)phosphoryl]oxy}methyl)-3,4-dihydroxyoxolan-2-yl]-3-carbamoyl-1λ⁵-pyridin-1-ylium
Synonyms: nadide sodium
Molecular Weight (g/mol): 685.41
Molecular Formula: C21H26N7NaO14P2
InChi Key: OGCURMAMSJFXSG-QYZPTAICSA-M
SMILES: [Na+].NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
Grade 
  • (4)
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)