accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COHOCl

CAS RN 2012-74-0

IUPAC Name: 2-(4-chlorophenyl)-3-methylbutanoic acid
Synonyms: 2-(4-chlorophenyl)-3-methylbutanoic acid
Molecular Weight (g/mol): 212.67
Molecular Formula: C11H13ClO2
InChi Key: VTJMSIIXXKNIDJ-UHFFFAOYNA-N
SMILES: CC(C)C(C(O)=O)C1=CC=C(Cl)C=C1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results