accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20129-11-7

IUPAC Name: 1-bromo-2,4,5-trimethoxybenzene
Synonyms: 1-bromo-2,4,5-trimethoxybenzene
Molecular Weight (g/mol): 247.09
Molecular Formula: C9H11BrO3
InChi Key: SFEPXIIFUHNCDO-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=C(OC)C=C1Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results