accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2013-12-9

IUPAC Name: (2R)-2-aminopentanoic acid
Synonyms: D-(-)-norvaline|D-ape|D-nva
Molecular Weight (g/mol): 117.15
Molecular Formula: C5H11NO2
InChi Key: SNDPXSYFESPGGJ-SCSAIBSYSA-N
SMILES: CCC[C@@H](N)C(O)=O
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)