accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClCl

CAS RN 2014-83-7

IUPAC Name: 1,3-dichloro-2-(chloromethyl)benzene
Synonyms: 1,3-dichloro-2-(chloromethyl)benzene
Molecular Weight (g/mol): 195.47
Molecular Formula: C7H5Cl3
InChi Key: LBOBESSDSGODDD-UHFFFAOYSA-N
SMILES: ClCC1=C(Cl)C=CC=C1Cl
Molecular Weight (g/mol) 
  • (3)
  • (1)
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)