accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 201485-17-8

IUPAC Name: (2S)-6-(carbamoylamino)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
Synonyms: (2S)-6-(carbamoylamino)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}hexanoic acid|FMOC-HOCIT-OH
Molecular Weight (g/mol): 411.46
Molecular Formula: C22H25N3O5
InChi Key: VJZUCXPETVPQIB-TWYLJJHKNA-N
SMILES: NC(=O)NCCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)