accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2015-56-7

IUPAC Name: {[amino(phenoxy)phosphoryl]oxy}benzene
Synonyms: {[amino(phenoxy)phosphoryl]oxy}benzene
Molecular Weight (g/mol): 249.21
Molecular Formula: C12H12NO3P
InChi Key: QWMUDOFWQWBHFI-UHFFFAOYSA-N
SMILES: NP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results