accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOHOOONH(R)H3CCH3

CAS RN 201531-88-6

IUPAC Name: (2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-methyl-3-[(triphenylmethyl)sulfanyl]butanoic acid
Synonyms: (2R)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-methyl-3-[(triphenylmethyl)sulfanyl]butanoic acid
Molecular Weight (g/mol): 613.77
Molecular Formula: C39H35NO4S
InChi Key: XSGMGAINOILNJR-PGUFJCEWSA-N
SMILES: CC(C)(SC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)