accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2016-42-4

IUPAC Name: hydrogen tetradecan-1-amine chloride
Synonyms: hydrogen tetradecylamine chloride
Molecular Weight (g/mol): 249.87
Molecular Formula: C14H32ClN
InChi Key: KKWUACQXLWHLCX-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCCCCCCCCCCCCCN
Boiling Point 
  • (2)
  • (5)
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)