accessibility menu, dialog, popup

UserName

Viewing profile as user:
Br

CAS RN 201731-79-5

IUPAC Name: 2-bromo-9,10-diphenylanthracene
Synonyms: 2-bromo-9,10-diphenylanthracene
Molecular Weight (g/mol): 409.33
Molecular Formula: C26H17Br
InChi Key: OZNXPZBQVNNJCS-UHFFFAOYSA-N
SMILES: BrC1=CC=C2C(=C1)C(C1=CC=CC=C1)=C1C=CC=CC1=C2C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results