accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OONNNOHO

CAS RN 201809-22-5

IUPAC Name: 6-{4-[(tert-butoxy)carbonyl]piperazin-1-yl}pyridine-3-carboxylic acid
Synonyms: 6-[4-(tert-butoxycarbonyl)piperazin-1-yl]pyridine-3-carboxylic acid
Molecular Weight (g/mol): 307.35
Molecular Formula: C15H21N3O4
InChi Key: PWYGTZUOLAGDNK-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCN(CC1)C1=NC=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results