accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3

CAS RN 20184-89-8

IUPAC Name: non-3-yne
Synonyms: 3-nonyne
Molecular Weight (g/mol): 124.23
Molecular Formula: C9H16
InChi Key: SRRDSRCWRHKEKU-UHFFFAOYSA-N
SMILES: CCCCCC#CCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results