accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 201852-49-5

IUPAC Name: potassium trifluoro(2-phenylethenyl)boranuide
Synonyms: potassium trifluoro(2-phenylethenyl)boranuide
Molecular Weight (g/mol): 210.05
Molecular Formula: C8H7BF3K
InChi Key: NONAUTDEFRJJII-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)C=CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results