accessibility menu, dialog, popup

UserName

Viewing profile as user:
II(E/Z)

CAS RN 201861-91-8

IUPAC Name: 1-iodo-4-[2-(4-iodophenyl)ethenyl]benzene
Synonyms: 1-iodo-4-[2-(4-iodophenyl)ethenyl]benzene
Molecular Weight (g/mol): 432.04
Molecular Formula: C14H10I2
InChi Key: BYEYAZGFUKIZPM-UHFFFAOYSA-N
SMILES: IC1=CC=C(C=CC2=CC=C(I)C=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)