accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20191-74-6

IUPAC Name: 2-ethyl-5-nitroaniline
Synonyms: 2-ethyl-5-nitroaniline
Molecular Weight (g/mol): 166.18
Molecular Formula: C8H10N2O2
InChi Key: MMZWMCKTKJKIMC-UHFFFAOYSA-N
SMILES: CCC1=C(N)C=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)