accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20232-39-7

IUPAC Name: 6,7-dimethoxy-3,4-dihydroisoquinoline hydrochloride
Synonyms: 6,7-dimethoxy-3,4-dihydroisoquinoline hydrochloride
Molecular Weight (g/mol): 227.69
Molecular Formula: C11H14ClNO2
InChi Key: PQXVEYYRJHMTEV-UHFFFAOYSA-N
SMILES: Cl.COC1=CC2=C(C=NCC2)C=C1OC
Percent Purity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)