accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 202414-68-4

IUPAC Name: N-[(4-chlorophenyl)(phenyl)methyl]formamide
Synonyms: N-[(4-chlorophenyl)(phenyl)methyl]formamide
Molecular Weight (g/mol): 245.71
Molecular Formula: C14H12ClNO
InChi Key: HDGOSEHHSVUCBO-UHFFFAOYNA-N
SMILES: ClC1=CC=C(C=C1)C(NC=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results