accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20265-35-4

IUPAC Name: 2-methoxy-3-nitropyridine
Synonyms: 2-methoxy-3-nitropyridine
Molecular Weight (g/mol): 154.13
Molecular Formula: C6H6N2O3
InChi Key: WZNQCVOSOCGWJG-UHFFFAOYSA-N
SMILES: COC1=NC=CC=C1[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results