accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20265-97-8

IUPAC Name: hydrogen 4-methoxyaniline chloride
Synonyms: hydrogen P-anisidine chloride
Molecular Weight (g/mol): 159.61
Molecular Formula: C7H10ClNO
InChi Key: VQYJLACQFYZHCO-UHFFFAOYSA-N
SMILES: [H+].[Cl-].COC1=CC=C(N)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (2)
  • (1)