CAS RN 20280-81-3
IUPAC Name:
4-methoxy-2H-chromen-2-one
Synonyms:
4-methoxychromen-2-one
Molecular Weight (g/mol):
176.17
Molecular Formula:
C10H8O3
InChi Key:
MLCMXDYMSAZNPC-UHFFFAOYSA-N
SMILES:
COC1=CC(=O)OC2=CC=CC=C12
1
–
1
of
1
results
1
Promotions Available
4-Methoxycoumarin, 98%
4-Methoxycoumarin, 98%
CAS: 20280-81-3 Molecular Formula: C10H8O3 Molecular Weight (g/mol): 176.171 MDL Number: MFCD00017349 InChI Key: MLCMXDYMSAZNPC-UHFFFAOYSA-N PubChem CID: 529426 IUPAC Name: 4-methoxychromen-2-one SMILES: COC1=CC(=O)OC2=CC=CC=C21