accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNClCl

CAS RN 202823-67-4

IUPAC Name: 3,6-dichloro-4,5-diazatricyclo[6.2.2.0²,⁷]dodeca-2(7),3,5-triene
Synonyms: 3,6-dichloro-4,5-diazatricyclo[6.2.2.0²,⁷]dodeca-2(7),3,5-triene
Molecular Weight (g/mol): 229.1
Molecular Formula: C10H10Cl2N2
InChi Key: GZIFYLGNTOAIJJ-UHFFFAOYSA-N
SMILES: ClC1=NN=C(Cl)C2=C1C1CCC2CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results