accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 202865-82-5

IUPAC Name: 1-(3-bromo-4-fluorophenyl)propan-1-one
Synonyms: 1-(3-bromo-4-fluorophenyl)propan-1-one
Molecular Weight (g/mol): 231.06
Molecular Formula: C9H8BrFO
InChi Key: IZSXDWBFACKNSW-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC(Br)=C(F)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results